Assay Details
| Gene Symbol: PLCL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027962005.1 | XP_027817806.1 | 2 - 3 | 2575 | 57 | |
| XM_027962015.1 | XP_027817816.1 | 2 - 3 | 2555 | 57 | ||
More Information
Set Membership: |
|

PLCL2
Genomic Map