Assay Details
| Gene Symbol: YAE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008003.4 | XP_004008052.2 | 2 - 3 | 287 | 100 | |
| XM_015095359.2 | XP_014950845.2 | 2 - 3 | 362 | 100 | ||
More Information
Set Membership: |
|

YAE1
Genomic Map