Assay Details
| Gene Symbol: ACER2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027964568.1 | XP_027820369.1 | 2 - 3 | 284 | 84 | |
| XM_027964569.1 | XP_027820370.1 | 2 - 3 | 284 | 84 | ||
| XM_027964570.1 | XP_027820371.1 | 2 - 3 | 284 | 84 | ||
More Information
Set Membership: |
|

ACER2
Genomic Map