Assay Details
| Gene Symbol: SMLR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012183324.3 | XP_012038714.2 | 2 - 3 | 200 | 72 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

SMLR1| Gene Symbol: SMLR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012183324.3 | XP_012038714.2 | 2 - 3 | 200 | 72 | |