Assay Details
| Gene Symbol: POFUT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004014475.4 | XP_004014524.1 | 1 - 2 | 218 | 65 | |
| XM_012189028.3 | XP_012044418.1 | 1 - 2 | 206 | 65 | ||
More Information
Set Membership: |
|

POFUT1
Genomic Map