Assay Details
| Gene Symbol: LOC101102156 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012486.3 | XP_004012535.2 | 8 - 9 | 732 | 96 | |
| XM_004012487.3 | XP_004012536.2 | 9 - 10 | 828 | 96 | ||
More Information
Set Membership: |
|

LOC101102156
Genomic Map