Assay Details
Gene Symbol: LOC101108520 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017934.4 | XP_004017983.1 | 4 - 5 | 758 | 80 | |
XM_004017935.4 | XP_004017984.1 | 4 - 5 | 720 | 80 | ||
XM_012098909.3 | XP_011954299.1 | 4 - 5 | 718 | 80 | ||
XM_012098910.3 | XP_011954300.1 | 4 - 5 | 589 | 80 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
