Assay Details
| Gene Symbol: SQOR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004010645.4 | XP_004010694.1 | 4 - 5 | 570 | 112 | |
| XM_012181644.3 | XP_012037034.1 | 4 - 5 | 543 | 112 | ||
More Information
Set Membership: |
|

SQOR
Genomic Map