Assay Details
| Gene Symbol: LOC101107224 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012097572.2 | XP_011952962.2 | 15 - 16 | 1670 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101107224| Gene Symbol: LOC101107224 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012097572.2 | XP_011952962.2 | 15 - 16 | 1670 | 79 | |
Set Membership: |
|