Assay Details
| Gene Symbol: MOCOS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960805.1 | XP_027816606.1 | 6 - 7 | 1221 | 59 | |
| XM_027960806.1 | XP_027816607.1 | 6 - 7 | 1641 | 59 | ||
More Information
Set Membership: |
|

MOCOS
Genomic Map