Assay Details
| Gene Symbol: LOC114111341 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963108.1 | XP_027818909.1 | 10 - 11 | 1472 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC114111341| Gene Symbol: LOC114111341 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963108.1 | XP_027818909.1 | 10 - 11 | 1472 | 91 | |
Set Membership: |
|