Assay Details
| Gene Symbol: SEPHS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027976639.1 | XP_027832440.1 | 7 - 8 | 1141 | 60 | |
| XM_027976640.1 | XP_027832441.1 | 7 - 8 | 918 | 60 | ||
More Information
Set Membership: |
|

SEPHS1
Genomic Map