Assay Details
| Gene Symbol: TGS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012183714.3 | XP_012039104.1 | 5 - 6 | 1348 | 65 | |
| XM_012183717.3 | XP_012039107.1 | 5 - 6 | 1275 | 65 | ||
| XM_012183719.3 | XP_012039109.1 | 5 - 6 | 1267 | 65 | ||
More Information
Set Membership: |
|

TGS1
Genomic Map