Assay Details
| Gene Symbol: LOC101118990 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027956841.1 | XP_027812642.1 | 2 - 3 | 263 | 73 | |
| XM_027956842.1 | XP_027812643.1 | 2 - 3 | 263 | 73 | ||
| XM_027956843.1 | XP_027812644.1 | 2 - 3 | 277 | 73 | ||

LOC101118990
Genomic Map