Assay Details
| Gene Symbol: LOC101123613 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963136.1 | XP_027818937.1 | 14 - 15 | 1735 | 82 | |
| XM_027963137.1 | XP_027818938.1 | 14 - 15 | 1735 | 82 | ||
| XM_027963138.1 | XP_027818939.1 | 14 - 15 | 1723 | 82 | ||
| Gene Symbol: LOC114109733 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027959224.1 | XP_027815025.1 | 1 | 2473 | 82 | |
More Information
Set Membership: |
|

LOC101123613
Genomic Map