Assay Details
| Gene Symbol: LOC101123613 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963136.1 | XP_027818937.1 | 30 - 31 | 4058 | 73 | |
| XM_027963138.1 | XP_027818939.1 | 30 - 31 | 4046 | 73 | ||
More Information
Set Membership: |
|

LOC101123613
Genomic Map