Assay Details
| Gene Symbol: NEMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015093322.2 | XP_014948808.2 | 3 - 4 | 528 | 85 | |
| XM_027964705.1 | XP_027820506.1 | 3 - 4 | 3259 | 85 | ||
| XM_027964706.1 | XP_027820507.1 | 4 - 5 | 542 | 85 | ||

NEMP2
Genomic Map