Assay Details
| Gene Symbol: JPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973056.1 | XP_027828857.1 | 2 - 3 | 1409 | 70 | |
| XM_027973057.1 | XP_027828858.1 | 2 - 3 | 1409 | 70 | ||
| XM_027973058.1 | XP_027828859.1 | 2 - 3 | 1409 | 70 | ||
| XM_027973059.1 | XP_027828860.1 | 2 - 3 | 1409 | 70 | ||
| XM_027973060.1 | XP_027828861.1 | 2 - 3 | 1409 | 70 | ||
| XM_027973061.1 | XP_027828862.1 | 2 - 3 | 1410 | 70 | ||
More Information
Set Membership: |
|

JPH1
Genomic Map