Assay Details
| Gene Symbol: HDGF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027976536.1 | XP_027832337.1 | 4 - 5 | 805 | 82 | |
| XM_027976543.1 | XP_027832344.1 | 4 - 5 | 580 | 82 | ||
| XM_027976545.1 | XP_027832346.1 | 4 - 5 | 423 | 82 | ||
More Information
Set Membership: |
|

HDGF
Genomic Map