Assay Details
| Gene Symbol: LOC101118470 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027978332.1 | XP_027834133.1 | 4 - 5 | 1280 | 98 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101118470| Gene Symbol: LOC101118470 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027978332.1 | XP_027834133.1 | 4 - 5 | 1280 | 98 | |