Assay Details
| Gene Symbol: CCDC180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027964404.1 | XP_027820205.1 | 21 - 22 | 2984 | 60 | |
| XM_027964405.1 | XP_027820206.1 | 21 - 22 | 3147 | 60 | ||
| XM_027964406.1 | XP_027820207.1 | 19 - 20 | 2672 | 60 | ||
More Information
Set Membership: |
|

CCDC180
Genomic Map