Assay Details
| Gene Symbol: FAM168B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015093295.2 | XP_014948781.1 | 3 - 4 | 346 | 88 | |
| XM_015093296.2 | XP_014948782.1 | 3 - 4 | 329 | 88 | ||
| XM_015093298.2 | XP_014948784.1 | 3 - 4 | 276 | 88 | ||
| XM_027964664.1 | XP_027820465.1 | 3 - 4 | 234 | 88 | ||
More Information
Set Membership: |
|

FAM168B
Genomic Map