Assay Details
| Gene Symbol: C23H18orf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012103637.3 | XP_011959027.1 | 4 - 5 | 1169 | 133 | |
| XM_015103706.2 | XP_014959192.1 | 4 - 5 | 1393 | 133 | ||
| XM_027960883.1 | XP_027816684.1 | 3 - 4 | 1207 | 133 | ||
More Information
Set Membership: |
|

C23H18orf25
Genomic Map