Assay Details
| Gene Symbol: ZNF280D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015097014.2 | XP_014952500.1 | 9 - 10 | 1090 | 87 | |
| XM_015097015.2 | XP_014952501.1 | 8 - 9 | 941 | 87 | ||
| XM_015097018.2 | XP_014952504.1 | 7 - 8 | 827 | 87 | ||
| XM_015097020.2 | XP_014952506.1 | 6 - 7 | 781 | 87 | ||
| XM_015097021.2 | XP_014952507.1 | 7 - 8 | 895 | 87 | ||
| XM_027971801.1 | XP_027827602.1 | 8 - 9 | 941 | 87 | ||
| XM_027971802.1 | XP_027827603.1 | 8 - 9 | 941 | 87 | ||
More Information
Set Membership: |
|

ZNF280D
Genomic Map