Assay Details
| Gene Symbol: ST18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012183700.3 | XP_012039090.2 | 16 - 17 | 2823 | 63 | |
| XM_027972950.1 | XP_027828751.1 | 10 - 11 | 2278 | 63 | ||
| XM_027972951.1 | XP_027828752.1 | 17 - 18 | 2866 | 63 | ||
| XM_027972952.1 | XP_027828753.1 | 15 - 16 | 2744 | 63 | ||
More Information
Set Membership: |
|

ST18
Genomic Map