Assay Details
Gene Symbol: ADAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012185353.3 | XP_012040743.1 | 10 - 11 | 1187 | 64 | |
XM_027975273.1 | XP_027831074.1 | 9 - 10 | 1413 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
