Assay Details
| Gene Symbol: CAPZA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004007870.4 | XP_004007919.2 | 6 - 7 | 650 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CAPZA2| Gene Symbol: CAPZA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004007870.4 | XP_004007919.2 | 6 - 7 | 650 | 93 | |
Set Membership: |
|