Assay Details
| Gene Symbol: RBM39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004014533.3 | XP_004014582.2 | 10 - 11 | 1359 | 58 | |
| XM_004014534.3 | XP_004014583.2 | 10 - 11 | 1359 | 58 | ||
| XM_012189129.3 | XP_012044519.2 | 11 - 12 | 1425 | 58 | ||
| XM_012189130.3 | XP_012044520.2 | 11 - 12 | 1426 | 58 | ||
| XM_027977136.1 | XP_027832937.1 | 9 - 10 | 2483 | 58 | ||
| XM_027977137.1 | XP_027832938.1 | 11 - 12 | 1430 | 58 | ||
More Information
Set Membership: |
|

RBM39
Genomic Map