Assay Details
| Gene Symbol: ZHX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972931.1 | XP_027828732.1 | 3 - 4 | 3556 | 117 | |
| XM_027972932.1 | XP_027828733.1 | 4 - 5 | 3713 | 117 | ||
More Information
Set Membership: |
|

ZHX2
Genomic Map