Assay Details
| Gene Symbol: DENR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027956454.1 | XP_027812255.1 | 3 - 4 | 239 | 79 | |
| XM_027956455.1 | XP_027812256.1 | 3 - 4 | 143 | 79 | ||
More Information
Set Membership: |
|

DENR
Genomic Map