Assay Details
| Gene Symbol: CYP2A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015267.4 | XP_004015316.2 | 6 - 7 | 1015 | 63 | |
| GenBank mRNA | HQ263377.1 | - | 6 - 7 | 1000 | 63 | |
More Information
Set Membership: |
|

CYP2A6
Genomic Map