Assay Details
| Gene Symbol: VAPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012103583.3 | XP_011958973.1 | 3 - 4 | 290 | 69 | |
| XM_015103658.2 | XP_014959144.1 | 3 - 4 | 577 | 69 | ||
| XM_015103659.2 | XP_014959145.1 | 3 - 4 | 577 | 69 | ||
More Information
Set Membership: |
|

VAPA
Genomic Map