Assay Details
| Gene Symbol: IPMK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012102441.3 | XP_011957831.3 | 3 - 4 | 660 | 62 | |
| XM_027960153.1 | XP_027815954.1 | 3 - 4 | 662 | 62 | ||
More Information
Set Membership: |
|

IPMK
Genomic Map