Assay Details
| Gene Symbol: LOC101113487 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004007023.3 | XP_004007072.1 | 9 - 10 | 1039 | 66 | |
| XM_012175713.2 | XP_012031103.1 | 9 - 10 | 1036 | 66 | ||
| XM_012175714.2 | XP_012031104.1 | 9 - 10 | 1039 | 66 | ||
| XM_012175715.3 | XP_012031105.1 | 9 - 10 | 1032 | 66 | ||
More Information
Set Membership: |
|

LOC101113487
Genomic Map