Assay Details
| Gene Symbol: NAB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015093319.2 | XP_014948805.1 | 7 - 8 | 1621 | 87 | |
| XM_015093320.2 | XP_014948806.1 | 7 - 8 | 1618 | 87 | ||
More Information
Set Membership: |
|

NAB1
Genomic Map