Assay Details
| Gene Symbol: RFLNB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975157.1 | XP_027830958.1 | 2 - 3 | 383 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RFLNB| Gene Symbol: RFLNB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975157.1 | XP_027830958.1 | 2 - 3 | 383 | 62 | |
Set Membership: |
|