Assay Details
| Gene Symbol: LOC101106662 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004819.3 | XP_004004868.1 | 2 - 3 | 249 | 95 | |
| XM_027965079.1 | XP_027820880.1 | 2 - 3 | 165 | 95 | ||
| Gene Symbol: LOC114108659 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027965078.1 | XP_027820879.1 | 2 - 3 | 249 | 95 | |
More Information
Set Membership: |
|

LOC101106662
Genomic Map