Assay Details
| Gene Symbol: NUP50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004007043.4 | XP_004007092.2 | 7 - 8 | 1552 | 61 | |
| XM_027968202.1 | XP_027824003.1 | 7 - 8 | 1302 | 61 | ||
More Information
Set Membership: |
|

NUP50
Genomic Map