Assay Details
| Gene Symbol: LOC101116059 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972313.1 | XP_027828114.1 | 29 - 30 | 2992 | 135 | |
| XM_027972314.1 | XP_027828115.1 | 29 - 30 | 2815 | 135 | ||
| XM_027972315.1 | XP_027828116.1 | 28 - 29 | 2836 | 135 | ||
| XM_027972316.1 | XP_027828117.1 | 29 - 30 | 2994 | 135 | ||
| XM_027972317.1 | XP_027828118.1 | 28 - 29 | 2827 | 135 | ||
| XM_027972319.1 | XP_027828120.1 | 28 - 29 | 2758 | 135 | ||
More Information
Set Membership: |
|

LOC101116059
Genomic Map