Assay Details
| Gene Symbol: LOC101116059 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972313.1 | XP_027828114.1 | 9 - 10 | 1074 | 71 | |
| XM_027972314.1 | XP_027828115.1 | 9 - 10 | 897 | 71 | ||
| XM_027972315.1 | XP_027828116.1 | 9 - 10 | 924 | 71 | ||
| XM_027972316.1 | XP_027828117.1 | 9 - 10 | 1091 | 71 | ||
| XM_027972317.1 | XP_027828118.1 | 9 - 10 | 924 | 71 | ||
| XM_027972318.1 | XP_027828119.1 | 9 - 10 | 924 | 71 | ||
| XM_027972319.1 | XP_027828120.1 | 9 - 10 | 924 | 71 | ||
More Information
Set Membership: |
|

LOC101116059
Genomic Map