Assay Details
| Gene Symbol: C15H11orf42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004016215.4 | XP_004016264.2 | 2 - 3 | 961 | 95 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

C15H11orf42| Gene Symbol: C15H11orf42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004016215.4 | XP_004016264.2 | 2 - 3 | 961 | 95 | |