Assay Details
| Gene Symbol: LOC101122882 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027978752.1 | XP_027834553.1 | 3 - 4 | 410 | 76 | |
| XM_027978753.1 | XP_027834554.1 | 2 - 3 | 235 | 76 | ||
| Gene Symbol: RDH13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027978806.1 | XP_027834607.1 | 3 - 4 | 410 | 76 | |
More Information
Set Membership: |
|

LOC101122882
Genomic Map