Assay Details
| Gene Symbol: BEND2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963216.1 | XP_027819017.1 | 6 - 7 | 1014 | 100 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

BEND2| Gene Symbol: BEND2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963216.1 | XP_027819017.1 | 6 - 7 | 1014 | 100 | |