Assay Details
| Gene Symbol: LOC101119354 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002909.4 | XP_004002958.2 | 1 - 2 | 156 | 69 | |
| XM_027979919.1 | XP_027835720.1 | 1 - 2 | 164 | 69 | ||
More Information
Set Membership: |
|

LOC101119354
Genomic Map