Assay Details
| Gene Symbol: IQCA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015091828.2 | XP_014947314.2 | 4 - 5 | 913 | 57 | |
| XM_015091829.2 | XP_014947315.2 | 3 - 4 | 780 | 57 | ||
| XM_027967301.1 | XP_027823102.1 | 4 - 5 | 913 | 57 | ||
More Information
Set Membership: |
|

IQCA1
Genomic Map