Assay Details
Gene Symbol: LOC101104401 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027971072.1 | XP_027826873.1 | 2 - 3 | 360 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
