Assay Details
| Gene Symbol: CYRIA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012164859.3 | XP_012020249.1 | 11 - 12 | 1579 | 93 | |
| XM_015094234.2 | XP_014949720.1 | 11 - 12 | 1120 | 93 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CYRIA| Gene Symbol: CYRIA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012164859.3 | XP_012020249.1 | 11 - 12 | 1579 | 93 | |
| XM_015094234.2 | XP_014949720.1 | 11 - 12 | 1120 | 93 | ||