Assay Details
| Gene Symbol: WDR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012100081.3 | XP_011955471.2 | 3 - 4 | 2799 | 56 | |
| XM_012100082.3 | XP_011955472.2 | 3 - 4 | 2799 | 56 | ||
More Information
Set Membership: |
|

WDR6
Genomic Map