Assay Details
| Gene Symbol: PDZRN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015102331.2 | XP_014957817.2 | 7 - 8 | 1509 | 66 | |
| XM_027958161.1 | XP_027813962.1 | 5 - 6 | 2791 | 66 | ||
| XM_027958162.1 | XP_027813963.1 | 5 - 6 | 2649 | 66 | ||
More Information
Set Membership: |
|

PDZRN3
Genomic Map