Assay Details
| Gene Symbol: FUS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012104405.3 | XP_011959795.2 | 14 - 15 | 1581 | 55 | |
| XM_012104406.3 | XP_011959796.2 | 14 - 15 | 1578 | 55 | ||
More Information
Set Membership: |
|

FUS
Genomic Map